Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AC-0347A

Sigma-Aldrich

A-1-Antichymotrypsin (EP384) Rabbit Monoclonal Primary Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

antibody form

Ig fraction of antiserum

clone

monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:200

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

General description

Alpha 1-Antichymotrypsin is an acute phase glycoprotein that functions by inhibiting proteases secreted by neutrophils and mast cells during periods of inflammation. It is primarily produced by hepatocytes and locally expressed in histiocytes, including macrophages and Kupffer cells in the liver. Alpha 1-Antichymotrypsin is a useful marker for malignant fibrous histiocytomas to aid in differentiating from other soft-tissue tumors such as liposarcomas or Ewing′s sarcoma.

Quality


IVD

IVD

IVD

RUO

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and &lt0.1% Sodium Azide

Other Notes

For a copy of the IFU and CofA contact IVDorder@ABCAM.com

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service