Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A8597

Sigma-Aldrich

1-Amino-3,3-diethoxypropane

liquid, ≥97%

Synonym(s):

3-Aminopropionaldehyde diethylacetal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H17NO2
CAS Number:
Molecular Weight:
147.22
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Product Name

1-Amino-3,3-diethoxypropane, ≥97%, liquid

biological source

synthetic (organic)

Assay

≥97%

form

liquid

solubility

water: miscible

density

0.91 g/mL at 25 °C (lit.)

storage temp.

room temp

SMILES string

CCOC(CCN)OCC

InChI

1S/C7H17NO2/c1-3-9-7(5-6-8)10-4-2/h7H,3-6,8H2,1-2H3

InChI key

PXXMSHBZYAOHBD-UHFFFAOYSA-N

General description

1-Amino-3,3-diethoxypropane is an effective modulator of alginate and chitosan based hydrogels and effectively used for the delivery of bone marrow stromal cells (BMSCs) for regeneration of cartilage.

Application

1-Amino-3,3-diethoxypropane has been used:
  • with alginate in the preparation of hydrogel
  • in the generation of polyethylene glycol (PEG) polyethylenimine (PEI) polyplexes conjugate for transfection studies in hepatocellular carcinoma cell cultures
  • as a component of pH-sensitive PEG derivative for lipopolyplexes formation

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

172.4 °F - closed cup

Flash Point(C)

78 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

BMSCs laden injectable amino-diethoxypropane modified alginate-chitosan hydrogel for hyaline cartilage reconstruction
Wan W, et al.
Journal of Material Chemistry B: Materials for Biology and Medicine, 3(9), 1990-2005 (2015)
Amine-reactive pyridylhydrazone-based PEG reagents for pH-reversible PEI polyplex shielding
Fella C, et al.
European Journal of Pharmaceutical Sciences, 34(4-5), 309-320 (2008)
Massimiliano Magro et al.
Amino acids, 51(4), 679-690 (2019-02-07)
The preservation of enzymatic activity is a fundamental requirement for exploiting hybrid nano-bio-conjugates, and the control over protein-nanoparticle interactions, leading to stable and catalytically active hybrids, represents the key for designing new biosensing platforms. In this scenario, surface active maghemite
Caroline R Basso et al.
Talanta, 197, 482-490 (2019-02-18)
A new immunosensor using hybrid nanomaterials for the detection of dengue virus was demonstrated in this work. This immunosensor composed of nanoparticles of γ-Fe2O3(SAMN) modified with MPA- SAMN@MPA was characterized by FTIR spectroscopy, transmission electron microscopy,quartz crystal microbalance, UV-vis and
Pyridylhydrazone-based PEGylation for pH-reversible lipopolyplex shielding
Nie Y, et al.
Biomaterials, 32(3), 858-869 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service