Skip to Content
MilliporeSigma

A11100P

DMT-dA(bz) Pharmadite®

Synonym(s):

Adenosine, N-benzoyl-5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-deoxy-, 3′-[2-cyanoethyl N,N-bis(1-methylethyl)phosphoramidite], Adenosine, N-benzoyl-5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-deoxy-, 3′-[2-cyanoethyl bis(1-methylethyl)phosphoramidite], DMT-dA(bz) Phosphoramidite, N-benzoyl-5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-deoxyadenosine, 3′-[2-cyanoethyl N,N-bis(1-methylethyl)phosphoramidite], N6-Benzoyl-5′-O-(4,4′-dimethoxytrityl)-2′-deoxyadenosine-3′-O-[O-(2-cyanoethyl)-N,N′-diisopropylphosphoramidite]

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C47H52N7O7P
CAS Number:
Molecular Weight:
857.93
NACRES:
NA.51
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
eCl@ss:
32120505
Assay:
≥99.5% (reversed phase HPLC), >99.5% (31P-NMR)
Biological source:
non-animal source (no BSE/TSE risk)
Form:
powder or granules
Solubility:
soluble, clear
Storage temp.:
2-8°C
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

non-animal source (no BSE/TSE risk)

Quality Segment

type

for DNA synthesis

product line

Proligo Reagents

assay

≥99.5% (reversed phase HPLC), >99.5% (31P-NMR)

form

powder or granules

technique(s)

oligo synthesis: suitable

impurities

>= 170ppm, none detected (S/N <2.5), ≤0.1% single unspecified Imprity (reversed phase HPLC), ≤0.3% P(III) 100-169ppm (31P-NMR), ≤0.3% dA2 (reversed phase HPLC, DMT-dA(bz) cyanoethyl-H-posphonate), ≤0.3% dA3 (reversed phase HPLC, DMT-dA(bz) phosphoramidate), ≤0.30 wt. % water content (Karl Fischer), ≤0.5% P(V) -25-99ppm (31P-NMR), ≤3.0 wt. % residual solvent content

color

white to off-white

solubility

soluble, clear

suitability

conforms to structure for H-NMR, conforms to structure for LC-MS

nucleoside profile

base: deoxyadenosine
base protecting group: benzoyl
2' protecting group: none
5' protecting group: DMT
deprotection: standard

storage temp.

2-8°C

SMILES string

COc1ccc(cc1)C(OC[C@H]2O[C@H](C[C@@H]2OP(OCCC#N)N(C(C)C)C(C)C)n3cnc4c(NC(=O)c5ccccc5)ncnc34)(c6ccccc6)c7ccc(OC)cc7

InChI

1S/C47H52N7O7P/c1-32(2)54(33(3)4)62(59-27-13-26-48)61-40-28-42(53-31-51-43-44(49-30-50-45(43)53)52-46(55)34-14-9-7-10-15-34)60-41(40)29-58-47(35-16-11-8-12-17-35,36-18-22-38(56-5)23-19-36)37-20-24-39(57-6)25-21-37/h7-12,14-25,30-33,40-42H,13,27-29H2,1-6H3,(H,49,50,52,55)/t40-,41+,42+,62?/m0/s1

InChI key

GGDNKEQZFSTIMJ-AKWFTNRHSA-N

General description

DMT-dA(bz) Pharmadite® is a Pharmadite® product line that represents a new class of standard protected DNA phosphoramidites that are designed with highly controlled impurity profiles and exceptional overall purity. Pharmadite phosphoramidites are characterized by their exceptional purity and well-defined impurity profile. In particular, they are essentially free from contaminants that interfere in coupling reactions, such as other nucleosidic or non-nucleosidic phosphoramidites (P(III)-contaminants). All remaining impurities in Pharmadite phosphopramidites, if present, are identified, characterized, and quantified. The impact of any such defined impurities on the synthesis of oligonucleotides is well understood and has been shown to be insignificant.

Application

Pharmadite products fulfill all requirements of starting materials for themanufacture of active pharmaceutical ingredients as defined in the EMEA "Note for guidance on the chemistry of the new active substance," making them suitable building blocks for oligonucleotide drugs.

Other Notes

Pharmadite phosphoramidites are manufactured in scales of up to multi-hundred kilos, under certified ISO 9001 quality systems.

Legal Information

Pharmadite is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under DMT-dA(bz) Pharmadite®


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library