Skip to Content
MilliporeSigma

Key Documents

Skip To

59920C

EX-CELL® Antifoam

gamma irradiated

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.75
UNSPSC Code:
12352207
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

sterility

γ-irradiated

form

liquid

technique(s)

NMR: suitable, cell culture | mammalian: suitable, protein quantification: suitable

suitability

suitable for manufacturing use

application(s)

mRNA manufacturing

shipped in

ambient

Application

EX-CELL® ANTIFOAM is a USP Grade non-ionic emulsion (simethicone) designed for use in the pharmaceutical and veterinary biological industries to aid in the control of foaming typically associated with the use of culture medium in bioreactors.
Through formulation optimization, utilization of a quantitative assay and an understanding of the nonspecific binding of simethicone, SAFC is able to report the actual simethicone concentration of the gamma irradiated finished material. As a result of this development, SAFC offers an optimized formulation ensuring a consistent product.
This product is of non-animal origin formulated from a 30% Simethicone Emulsion from DOW CORNING Q7-2587

Packaging

  • 59920C-1B (1L): Packaged in a single-use polyethylene (PE) film bag, it includes two customer outlet tubings: one 24-inch C-Flex TPE and one 18-inch C-Flex TPE.
  • 59920C-1BC (1L): Packaged in a single-use polyethylene (PE) film bag, it includes two customer outlet tubings: one 24-inch M-Flex TPE and one 18-inch M-Flex TPE.
  • 59920C-5B (5L): Packaged in a single-use polyethylene (PE) film bag, it includes two customer outlet tubings: one 24-inch C-Flex TPE and one 18-inch C-Flex TPE.
  • 59920C-5BC (5L): Packaged in a single-use polyethylene (PE) film bag, it includes two customer outlet tubings: one 24-inch M-Flex TPE and one 18-inch M-Flex TPE.
  • 59920C-10BC (10L): Packaged in a single-use polyethylene (PE) film bag, it includes two customer outlet tubings: one 24-inch M-Flex TPE and one 18-inch M-Flex TPE.

Legal Information

EX-CELL is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
14650C8346540236
description

gamma irradiated

description

Animal-component free, Protein-free, Chemically defined, without L-glutamine, liquid, sterile-filtered, suitable for cell culture

description

concentrate

description

~30% in H2O

form

liquid

form

liquid

form

liquid

form

liquid

technique(s)

protein quantification: suitable, cell culture | mammalian: suitable, NMR: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

protein quantification: suitable, HPLC: suitable

technique(s)

protein quantification: suitable

sterility

γ-irradiated

sterility

sterile-filtered

sterility

-

sterility

-

shipped in

ambient

shipped in

ambient

shipped in

-

shipped in

-

suitability

suitable for manufacturing use

suitability

-

suitability

-

suitability

-

Quality Level

500

Quality Level

400

Quality Level

100

Quality Level

200


Storage Class

12 - Non Combustible Liquids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.



Questions

Reviews

No rating value

Active Filters