Skip to Content
MilliporeSigma

47190

5-Fluoroorotic acid hydrate

≥99.0% (TLC)

Synonym(s):

2,6-Dihydroxy-5-fluoropyrimidine-4-carboxylic acid, 5-Fluorouracil-4-carboxylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C5H3FN2O4 · xH2O
CAS Number:
Molecular Weight:
174.09 (anhydrous basis)
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12352204
EC Number:
211-876-0
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥99.0% (TLC)

form

solid

mp

278 °C (dec.) (lit.)

solubility

4 M NH4OH: 50 mg/mL, clear, faintly yellow

storage temp.

−20°C

SMILES string

OC(C(NC(N1)=O)=C(F)C1=O)=O.[H]O[H]

InChI

1S/C5H3FN2O4.H2O/c6-1-2(4(10)11)7-5(12)8-3(1)9;/h(H,10,11)(H2,7,8,9,12);1H2

InChI key

LODRRYMGPWQCTR-UHFFFAOYSA-N

Application

Useful in the selection of orotidine-5′-phosphate decarboxylase mutants of Saccharomyces cerevisiae.

Other Notes

Selective agent in yeast molecular genetics


Still not finding the right product?

Explore all of our products under 5-Fluoroorotic acid hydrate


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library