Skip to Content
MilliporeSigma
5-Fluoroorotic acid hydrate ≥99.0% (TLC)

5-Fluoroorotic acid hydrate

5-Fluoroorotic acid hydrate ≥99.0% (TLC)
Synonym(s):
2,6-Dihydroxy-5-fluoropyrimidine-4-carboxylic acid, 5-Fluorouracil-4-carboxylic acid
Empirical Formula (Hill Notation):
C5H3FN2O4 · xH2O
CAS Number:
Molecular Weight:
174.09 (anhydrous basis)
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing