Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

45405

Sigma-Aldrich

(±)-Equol

≥99.0% (TLC)

Synonym(s):

3,4-Dihydro-3-(4-hydroxyphenyl)-2H-1-benzopyran-7-ol, 4′,7-Dihydroxyisoflavane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H14O3
CAS Number:
Molecular Weight:
242.27
Beilstein:
87752
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥99.0% (TLC)

form

solid

SMILES string

Oc1ccc(cc1)C2COc3cc(O)ccc3C2

InChI

1S/C15H14O3/c16-13-4-1-10(2-5-13)12-7-11-3-6-14(17)8-15(11)18-9-12/h1-6,8,12,16-17H,7,9H2

InChI key

ADFCQWZHKCXPAJ-UHFFFAOYSA-N

Gene Information

rat ... Ar(24208)

Biochem/physiol Actions

Metabolite of daidzein. Weak estrogenic agonist, and competitive inhibitor of 17β-estradiol at the estradiol receptor.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Joseph T Reed et al.
Cardiovascular research, 116(7), 1372-1385 (2019-11-19)
The myogenic reactivity of the middle cerebral arteries (MCA) protects the brain by altering the diameter in response to changes in lumen pressure. Large conductance potassium (BK) channels are known to regulate the myogenic reactivity, yet, it is not clear
Scott M Belcher et al.
BMC pharmacology & toxicology, 18(1), 63-63 (2017-09-08)
Our previous studies demonstrated that growth and migration of medulloblastoma (MB), the most common malignant brain tumor in children, are stimulated by 17β-estradiol. The growth stimulating effects of estrogens are mediated through ERβ and insulin-like growth factor 1 signaling to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service