Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C7H6Cl2N2O · 0.5ZnCl2
CAS Number:
Molecular Weight:
273.19
NACRES:
NA.47
PubChem Substance ID:
UNSPSC Code:
12171500
EC Number:
268-183-1
MDL number:
Product Name
Fast Red RC Salt,
form
powder
Quality Segment
composition
Dye content, ~20%
color
faint yellow to yellow
solubility
water: 1 mg/mL
application(s)
diagnostic assay manufacturing
hematology
histology
storage temp.
room temp
SMILES string
[Cl-].[Cl-].Cl[Zn]Cl.COc1ccc(Cl)cc1[N+]#N.COc2ccc(Cl)cc2[N+]#N
InChI
1S/2C7H6ClN2O.4ClH.Zn/c2*1-11-7-3-2-5(8)4-6(7)10-9;;;;;/h2*2-4H,1H3;4*1H;/q2*+1;;;;;+2/p-4
InChI key
YHUGASVQWCSKGW-UHFFFAOYSA-J
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| F2256-25G | 04061833612866 |