Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

42385

Sigma-Aldrich

Kollicoat® MAE 30 DP

46.0-50.6% methacrylic acid basis (calc. on solids content)

Synonym(s):

Methacrylic acid copolymer dispersion, Methacrylic acid-ethyl acrylate copolymer (1:1) dispersion 30 %

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.25

Quality Level

concentration

46.0-50.6% (methacrylic acid, calc. on solids content)

impurities

≤1 ppm arsenic (verified on random samples only)
≤1.00% coagulum content /particulate matter
≤10 ppm heavy metals (verified on random samples only)

ign. residue

≤0.1%

evapn. residue (140 °C, 30 min.)

28.5-31.5%

loss

68.5-71.5% loss on drying, 140°C, 30 min.

pH

2.1-3.0

viscosity

≤15 mPa.s(20 °C) (apparent viscosity)
3-15(20 °C) (mm2/s)

density

1.062-1.072

SMILES string

O(CC)C(=O)C=C.OC(=O)C(=C)C

InChI

1S/C5H8O2.C4H6O2/c1-3-5(6)7-4-2;1-3(2)4(5)6/h3H,1,4H2,2H3;1H2,2H3,(H,5,6)

InChI key

GDCRSXZBSIRSFR-UHFFFAOYSA-N

Related Categories

Application

Kollicoat polymers can be employed as film coatings (intelligent surfaces) with controlled-release agents, for instant-release or sustained-release applications. Kollicoat polymers can be used with all standard coating equipment and are cost-effective, delivering maximum quality in terms of function, stability, and appearance. This research grade product is intended for use in R&D and development only.

Analysis Note

appearance of a film : clear

Legal Information

Kollicoat is a registered trademark of BASF SE

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 4

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

A Dashevsky et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 58(1), 45-49 (2004-06-23)
The objective of this study was to obtain pH-independent release profiles from coated pellets containing drugs with pH-dependent solubility. pH-independent release of the basic model drug verapamil HCl was achieved by coating with a combination of the neutral polymer dispersions
Comparative studies with Kollicoat MAE 30 D and Kollicoat MAE 30 DP in aqueous spray dispersions and enteric coatings on highly swellable caffeine cores.
Dangel C, Schepky G, Reich HB, Kolter K.
Drug Devel. Ind. Pharm., 26, 415-421 (2000)
C Guthmann et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 69(2), 667-674 (2008-01-30)
The drug substance SAG/ZK has a short biological half-life and because of its weakly basic nature a strong pH-dependent solubility was observed. The aim of this study was to develop a controlled release (cr) multiple unit pellet formulation for SAG/ZK
H Kranz et al.
International journal of pharmaceutics, 380(1-2), 112-119 (2009-07-28)
The major aim of this study was to identify an efficient tool to adjust drug release patterns from aqueous and organic ethylcellulose (a gastrointestinal insoluble polymer) coated pellets and to evaluate the long term stability of the film coatings. Drug

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service