Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

417D-250

Sigma-Aldrich

Human Osteoblast Differentiation Medium (250 ml)

Synonym(s):

Osteogenic medium

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.71

form

liquid

Quality Level

shelf life

6 mo.

manufacturer/tradename

Cell Applications, Inc

shipped in

wet ice

storage temp.

2-8°C

General description

Human Osteoblast Differentiation Medium

Application

Human Osteoblast Differentiation Medium (250 ml) has been used to induce differentiation in human primary osteoblasts.

Quality

Each lot was tested for the ability to support plating, spreading and proliferation

Preparation Note

contained in protocol

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Convex vs concave surface nano-curvature of Ta2O5 thin films for tailoring the osteoblast adhesion
Vaidulych M, et al.
Surface and Coatings Technology, 393, 125805-125805 (2020)

Protocols

Store the cryovials in a liquid nitrogen storage tank immediately upon arrival.

Store the cryovials in a liquid nitrogen storage tank immediately upon arrival.

Store the cryovials in a liquid nitrogen storage tank immediately upon arrival.

Store the cryovials in a liquid nitrogen storage tank immediately upon arrival.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service