Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

311D-250

Sigma-Aldrich

Smooth Muscle Cell Differentiation Medium (250 ml)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.71

form

liquid

Quality Level

shelf life

6 mo.

manufacturer/tradename

Cell Applications, Inc

shipped in

wet ice

storage temp.

2-8°C

General description

Smooth Muscle Cell Differentiation Medium

Quality

Each lot was tested for the ability to support plating, spreading and proliferation

Preparation Note

contained in protocol

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Technical information for working with human coronary artery smooth muscle cells including thawing, subculturing and cryopreservation

Technical information for working with human coronary artery smooth muscle cells including thawing, subculturing and cryopreservation

Technical information for working with human coronary artery smooth muscle cells including thawing, subculturing and cryopreservation

Technical information for working with human coronary artery smooth muscle cells including thawing, subculturing and cryopreservation

Protocols

Technical information for working with human pulmonary artery smooth muscle cells including thawing, subculturing and cryopreservation

Technical information for working with human pulmonary artery smooth muscle cells including thawing, subculturing and cryopreservation

Technical information for working with human pulmonary artery smooth muscle cells including thawing, subculturing and cryopreservation

Technical information for working with human pulmonary artery smooth muscle cells including thawing, subculturing and cryopreservation

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service