Skip to Content

C-22062

Smooth Muscle Cell Growth Medium 2

Ready-to-use kit including Basal Medium and SupplementMix, 500 ml

Synonym(s):

Smooth Muscle Cell Medium, Smooth Muscle Growth Medium

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.71
UNSPSC Code:
12352207
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

liquid

packaging

pkg of 500 ml

technique(s)

cell culture | mammalian: suitable

shipped in

wet ice

storage temp.

2-8°C

General description

Smooth Muscle Cell Growth Medium 2 (Ready-to-use)

Application

Smooth Muscle Cell Growth Medium 2 is a low-serum (5% V/V) medium developed to establish and maintain smooth muscle cell cultures from blood vessels and hollow organs. The medium is optimized for primary human cells but can also be used for bovine, porcine, murine, dog, and rat smooth muscle cells.Recommended for:Human Umbilical Artery Smooth Muscle Cells (HUASMC)Human Coronary Artery Smooth Muscle Cells (HCASMC)Human Pulmonary Artery Smooth Muscle Cells (HPASMC)Human Aortic Smooth Muscle Cells (HAoSMC)Human Bronchial Smooth Muscle Cells (HBSMC)Human Tracheal Smooth Muscle Cells (HTSMC)Human Uterine Smooth Muscle Cells (HUtSMC)

Packaging

Part of a Cell Culture System offered in a variety of formats:Growth Medium (Ready-to-use): Includes Basal Medium and SupplementMixGrowth Medium Kit: Includes Basal Medium and SupplementPackSupplementMix: Contains all media supplements premixed in one vialSupplementPack: Contains all media supplements as individual vials

Preparation Note

Click here for more information.
Final supplement concentrations after addition to the basal medium:Fetal Calf Serum: 0.05 ml / mlEpidermal Growth Factor (recombinant human): 0.5 ng / mlBasic Fibroblast Growth Factor (recombinant human): 2 ng / mlInsulin (recombinant human): 5 µg / ml
Store the Basal Medium at 4 to 8°C in the dark and the SupplementMix or SupplementPack at -20°C immediately after arrival. Do not freeze the Basal Medium. If stored properly, the products are stable until the expiry date stated on the label. After adding the supplements to the Basal Medium, the shelf life of the complete medium is 6 weeks at 4 to 8°C. Do not freeze the complete medium. For use, pre-warm only an aliquot of the complete medium and keep the remaining medium refrigerated at 4 to 8°C.

Analysis Note

All lots of PromoCell Smooth Muscle Cell Growth Medium are subjected to comprehensive quality control tests using primary human smooth muscle cells. Each lot is checked for growth promoting activity, adherence rate, and typical morphology of the tested smooth muscle cells. Approved in-house lots of media are used as a reference. In addition, all lots of media have been tested for the absence of microbial contaminants (fungi, bacteria, mycoplasma).


Still not finding the right product?

Explore all of our products under Smooth Muscle Cell Growth Medium 2


Storage Class

12 - Non Combustible Liquids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library