Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

30326

Sigma-Aldrich

EMPIGEN® BB detergent

~30% active substance

Synonym(s):

N,N-Dimethyl-N-dodecylglycine betaine, N-(Alkyl C10-C16)-N,N-dimethylglycine betaine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)8-14CH2N+(CH3)2CH2COO-
CAS Number:
Beilstein:
3670807
EC Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.25

description

zwitterionic

Quality Level

form

liquid

composition

solid content, 35.0-45.0 % (w/v)

concentration

~30% active substance

CMC

1.6-2.1 mM (20-25°C)

density

1.030-1.060 g/mL at 20 °C

UV absorption

λ: 417 nm Amax: ≤0.055
λ: 441 nm Amax: ≤0.037

SMILES string

CCCCCCCCCCCC[N+](C)(C)CC([O-])=O

InChI

1S/C16H33NO2/c1-4-5-6-7-8-9-10-11-12-13-14-17(2,3)15-16(18)19/h4-15H2,1-3H3

InChI key

DVEKCXOJTLDBFE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Empigen BB is a zwitterionic detergent compatible with anionic, non-ionic, cationic and other amphoteric surfactants.

Empigen BB is a useful detergent that has been used in the solubilization of membrane proteins and biochemical analysis of keratins. It not only helps to solubilize bigger amounts of protein but it also maintains antibody reactivity. It has been also used in the purification of leucine-rich repeat kinase 2 (LRRK2) in Escherichia coli during research in Parkinson′s disease. The use of Empigen in the preparation of viral antigens is being increasing due to its effectivity in giving greater antibody responses.

Legal Information

EMPIGEN is a registered trademark of Huntsman Corporation

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Rinke et al.
Virology, 170(2), 424-432 (1989-06-01)
We have established a sensitive assay for the quantitative determination of large T antigen determinants on the surface of living simian virus 40 (SV40)-transformed cells (mKSA). Cells in suspension culture were incubated with monoclonal antibodies specific for large T antigen
Islam Khan et al.
Microbes and infection, 7(5-6), 853-859 (2005-05-10)
Campylobacter jejuni is a major food-borne pathogen and a leading cause of diarrhoea. A cytotoxin is most likely involved in the pathogenesis of inflammatory diarrhoea due to C. jejuni. A 45-kDa outer membrane protein encoded by the porA gene was
Wenjin Guo et al.
Protein expression and purification, 90(1), 34-39 (2013-05-15)
Vaccinia virus (VV) has been used to express a variety of heterologous proteins, including HIV envelope (Env) glycoproteins. The Env protein is synthesized as a precursor molecule, gp160, which is cleaved into the surface antigen gp120 and the transmembrane protein
James K Wahl
Hybridoma and hybridomics, 21(1), 37-44 (2002-05-07)
Desmosomes are the most prominent cell-cell junctions in most epithelial cells and serve to link the intermediate filament cytoskeletons of adjacent cells. Desmogleins and desmocollins are the transmembrane core of the desmosome and both are members of the cadherin family
Kulbir Singh et al.
Langmuir : the ACS journal of surfaces and colloids, 30(13), 3673-3680 (2014-03-15)
In this article, we report additive-induced micellar growth and rheology modification for mixtures of anionic (sodium dodecyl sulfate, SDS) and zwitterionic (N-alkylated glycine derivative, Empigen BB or EBB) surfactants. Two nonaromatic hydrotropic salts (hexyltrimethylammonium bromide, C6TAB, and/or dibutylenebis(dimethylbutylammonium bromide), 4-4-4)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service