Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

267R-1

Sigma-Aldrich

IgA (EP170) Rabbit Monoclonal Primary Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

EP170, monoclonal

description

(For In Vitro Diagnostic Use in Select Regions (See Chart))

form

buffered aqueous solution

species reactivity

human

packaging

bottle of 1.0 mL predilute (267R-17)
bottle of 7.0 mL predilute (267R-18)
vial of 0.1 mL concentrate (267R-14)
vial of 0.5 mL concentrate (267R-15)
vial of 1.0 mL concentrate (267R-16)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

isotype

IgG

shipped in

wet ice

storage temp.

2-8°C

General description

Immunoglobulin A (IgA) plays a critical role in mucosal immunity. It is present in the mucosal secretions such as tears, saliva, colostrum, intestinal juice, vaginal fluid, and secretions from the prostate and respiratory epithelium, and represents a key first line of defense against invasion by inhaled and ingested pathogens at the vulnerable mucosal surfaces. It is also found in small amounts in blood. Because it is resistant to degradation by enzymes, secretory IgA can survive in harsh environments such as the digestive and respiratory tracts, to provide protection against microbes that multiply in body secretions. It is useful when identifying multiple myeloma and plasmacytomas.1-3

Quality


IVD

IVD

IVD

RUO

Linkage

IgA (polyclonal) Positive Control Slides, Product No. 267S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide.

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nadeem A Ansari et al.
Asian Pacific journal of cancer prevention : APJCP, 8(4), 593-596 (2008-02-12)
The frequency of expression of immunoglobulin (Ig) light and heavy chain isotypes was analyzed in myeloma proteins (M-proteins) from sera of 40 Indian patients with clinically established multiple myeloma. Patients samples were screened by a combination of electrophoresis, immunoelectrophoresis (IEP)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service