Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP0543

Sigma-Aldrich

LAG3 (CD223), Biotin Labeled

recombinant, expressed in HEK 293 cells

Synonym(s):

CD223, Lag, Lag-3, Lymphocyte-Activation Gene 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.26

recombinant

expressed in HEK 293 cells

form

frozen liquid

mol wt

74.8 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... LAG3(3902)

General description

Human secreted LAG3, Fc fusion protein, also known as Lymphocyte-Activation Gene 3 and CD223, with C-terminal Avitag. GenBank Accession No. NM_002286, a.a. 23-450 expressed in a HEK293 cell expression system and enzymatically biotin-labeled using Avitag technology. MW = 74.8 kDa (monomer). This protein runs at a higher MW by SDS-PAGE due to glycosylation.

Physical form

Aqueous solution

Legal Information

AviTag is a trademark of Avidity, LLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service