Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

207A-7

Sigma-Aldrich

FSH Rabbit Polyclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

Ig fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (207A-74)
vial of 0.5 mL concentrate (207A-75)
bottle of 1.0 mL predilute (207A-77)
vial of 1.0 mL concentrate (207A-76)
bottle of 7.0 mL predilute (207A-78)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:25-1:100

control

pituitary

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

General description

Follicle-stimulating hormone (FSH) is a member of the pituitary glycoprotein hormone family which includes luteinizing hormone, chorionic gonadotropin, and thyroid-stimulating hormone. Follicle-stimulating hormone enables ovarian folliculogenesis to the antral follicle stage and is essential for Sertoli cell proliferation and maintenance of sperm quality in the testis. Members of the pituitary glycoprotein hormone family consist of a shared alpha chain, and a beta chain encoded by a separate gene. The FSHB gene encodes the beta subunit of follicle stimulating hormone. Anti-FSH is a useful marker in classification of pituitary tumors and the study of pituitary disease, as it reacts with FSH-producing cells (gonadotrophs).

Quality


IVD

IVD

IVD

RUO

Linkage

FSH Positive Control Slides, Product No. 207S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Schmid et al.
Pathology, research and practice, 197(10), 663-669 (2001-11-10)
Null cell adenomas and oncocytomas are clinically inactive adenomas of the pituitary gland. They do not show any significant hormone content detectable by immunohistochemistry. This study aimed at demonstrating mRNAs for all main pituitary hormones in 32 null cell adenomas
S Uccella et al.
Pituitary, 3(3), 131-139 (2001-06-01)
The localization of inhibin/activin (I/A) subunits was investigated in human normal adenohypophysial cells and in 87 pituitary adenomas of different types, using immunohistochemistry. Monoclonal antibodies directed against alpha, beta A and beta B subunits of I/A were employed. In normal
S La Rosa et al.
Virchows Archiv : an international journal of pathology, 437(3), 264-269 (2000-10-19)
Gonadotropin-releasing hormone (GnRH), which is a well-known regulator of gonadotroph function, has recently been considered to be a paracrine factor involved in the control of somatotroph, lactotroph, and corticotroph cells. GnRH action is initiated by binding to a specific cell

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service