Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

19511

Sigma-Aldrich

Mant-ADP triethylammonium salt solution

≥95.0% (HPLC)

Synonym(s):

2′/3′-O-(N-Methylanthraniloyl)adenosine 5′-diphosphate triethylammonium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H22N6O11P2 · xC6H15N
CAS Number:
Molecular Weight:
560.35 (free acid basis)
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥95.0% (HPLC)

form

liquid

pH

7.0-8.0

storage temp.

−20°C

SMILES string

OP(O)(OP(O)(OC[C@@H]1[C@@H]([R1])[C@@H]([R2])[C@H](N(C=N2)C3=C2C(N)=NC=N3)O1)=O)=O.CCN(CC)CC.O=C(C4=C(NC)C=CC=C4)O.O=C(C5=C(NC)C=CC=C5)O

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service