Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

14324C

SAFC

EX-CELL® 302 Serum-Free Medium for CHO Cells

without L-glutamine, liquid, sterile-filtered, suitable for cell culture

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75

description

for research or for further manufacturing use

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

components

L-glutamine: no
NaHCO3: 1.60 g/L
HEPES: 7.5 mM
glucose: 3.42 g/L
phenol red: yes

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

EX-CELL 302 is a serum-free medium which has been specifically developed for the long-term growth of transformed Chinese Hamster Ovary (CHO) cells in suspension for the expression of antibodies or protein products. EX-CELL 302 is an appropriate medium for use with the DHFR- or Glutamine Synthetase, or the GS System, because it does not contain hypoxanthine, thymidine or L-glutamine. For use with the DHFR- selection system we recommend the addition of 4 mM L-glutamine for optimal growth. Due to current regulatory concerns about the sources of raw materials, EX-CELL 302 was developed using only recombinant human proteins that have molecular weights less than 10 kDa. The total protein concentration found in EX-CELL 302 is less than 1 mg/L. Lutrol has been added at a final concentration of 0.1% to EX-CELL 302 to protect against shear damage in sparged bioreactor systems.

Caution

Do not freeze. Protect from light. Several components of the medium are light labile and should not be exposed to light for lengthy periods of time.

Legal Information

EX-CELL is a registered trademark of Merck KGaA, Darmstadt, Germany
GS System is a trademark of Lonza Group Ltd.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service