Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

14360C

SAFC

EX-CELL® CD CHO Serum-Free Medium for CHO Cells, Chemically Defined

Animal-component free, without L-glutamine, without hypoxanthine, without thymidine, liquid, sterile-filtered, suitable for cell culture

Pharma Manufacturing

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75

Quality Level

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

components

phenol red: yes
L-glutamine: no
NaHCO3: 1.6 g/L
glucose: 5.6 g/L
HEPES: 7.5 mM

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

EX-CELL CD CHO is a chemically defined, animal-component free, serum-free medium developed for the long-term growth of Chinese Hamster Ovary (CHO) cells and expression of antibodies or protein products in suspension cell culture. CHO suspension cultures can be subcultured directly into EX-CELL CD CHO from serum-supplemented or serum-free medium with little or no adaptation. EX-CELL CD CHO is formulated without hypoxanthine, thymidine or L-glutamine, making it an appropriate medium for selection systems such as DHFR- and Glutamine Synthetase (GS System™). For applications that do not require the selective pressure of a hypoxanthine/thymidine (HT)-deficient medium, we recommend the use of EX-CELL CD CHO with hypoxanthine and thymidine (Catalog No. 14361C).

Legal Information

EX-CELL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

MGAT1 adds N-acetylglucosamine to the Man5GlcNAc2 (Man5) structure. Goh et al. reported increased sialylation after restoring MGAT1 function in MGAT1 deficient CHO cells.

A signal peptide is a 5-30 amino acid (aa) peptide present at the N-terminus of secretory proteins.

In the present study, we have identifi ed species-specifi c housekeeping genes (HKGs) for Chinese Hamster Ovary (CHO) cells using data from microarray gene expression profiling.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service