Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C-23010

Sigma-Aldrich

Fibroblast Growth Medium

Ready-to-use kit including Basal Medium and SupplementMix, 500 ml

Synonym(s):

Primary Cell Culture Media

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.71

form

aqueous solution

packaging

pkg of 500 ml

technique(s)

cell culture | mammalian: suitable

shipped in

wet ice

General description

Fibroblast Growth Medium (Ready-to-use)

Application

Fibroblast Growth Medium is optimized for the cultivation of neonatal and juvenile fibroblasts. While the supplements of the Fibroblast Growth Medium are serum-free, the Basal Medium is manufactured with 0.5% serum. The medium is optimized for primary human cells, but can also be used for bovine, porcine, and rat fibroblasts as well as fibroblast cell lines.Recommended for:Normal Human Dermal Fibroblasts (NHDF), juvenile foreskin

Components

Part of a Cell Culture System offered in a variety of formats:Growth Medium (Ready-to-use): Includes Basal Medium and SupplementMixGrowth Medium Kit: Includes Basal Medium and SupplementPackSupplementMix: Contains all media supplements premixed in one vialSupplementPack: Contains all media supplements as individual vials

Quality

All lots of PromoCell Fibroblast Growth Medium are subjected to comprehensive quality control tests using primary human fibroblasts. Each lot is checked for growth promoting activity, adherence rate, and typical morphology of the tested fibroblasts. Approved in-house lots of media are used as a reference. In addition, all lots of media have been tested for the absence of microbial contaminants (fungi, bacteria, mycoplasma).

Preparation Note

Final supplement concentrations after addition to the basal medium:Basic Fibroblast Growth Factor (recombinant human): 1 ng / ml Insulin: 5 µg / ml

Storage and Stability

Store the Basal Medium at 4 to 8°C in the dark and the SupplementMix or SupplementPack at -20°C immediately after arrival. Do not freeze the Basal Medium. If stored properly, the products are stable until the expiry date stated on the label. After adding the supplements to the Basal Medium, the shelf life of the complete medium is 6 weeks at 4 to 8°C. Do not freeze the complete medium. For use, pre-warm only an aliquot of the complete medium and keep the remaining medium refrigerated at 4 to 8°C.

Subculture Routine

Click here for more information.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Primary human fibroblasts and growth medium for the in vitro cultivation of juvenile and adult fibroblasts. Protocols for handling, cell passaging, media, and product use.

Primary human fibroblasts and growth medium for the in vitro cultivation of juvenile and adult fibroblasts. Protocols for handling, cell passaging, media, and product use.

Primary human fibroblasts and growth medium for the in vitro cultivation of juvenile and adult fibroblasts. Protocols for handling, cell passaging, media, and product use.

Primary human fibroblasts and growth medium for the in vitro cultivation of juvenile and adult fibroblasts. Protocols for handling, cell passaging, media, and product use.

Related Content

Explore expert protocols and culture media for human primary cell lines to replicate biological systems accurately.

Explore expert protocols and culture media for human primary cell lines to replicate biological systems accurately.

Explore expert protocols and culture media for human primary cell lines to replicate biological systems accurately.

Explore expert protocols and culture media for human primary cell lines to replicate biological systems accurately.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service