Skip to Content
MilliporeSigma
All Photos(1)

Documents

P52652

Sigma-Aldrich

Propylene carbonate

ReagentPlus®, 99%

Synonym(s):

1,2-Propanediol cyclic carbonate, 4-Methyl-1,3-dioxolan-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6O3
CAS Number:
Molecular Weight:
102.09
Beilstein:
107913
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.21

vapor pressure

0.13 mmHg ( 20 °C)
0.98 mmHg ( 50 °C)

Quality Level

product line

ReagentPlus®

Assay

99%

form

liquid

autoignition temp.

851 °F

expl. lim.

14.3 %

refractive index

n20/D 1.421 (lit.)

pH

7 (20 °C, 200 g/L)

bp

240 °C (lit.)

mp

−55 °C (lit.)

density

1.204 g/mL at 25 °C (lit.)

SMILES string

CC1COC(=O)O1

InChI

1S/C4H6O3/c1-3-2-6-4(5)7-3/h3H,2H2,1H3

InChI key

RUOJZAUFBMNUDX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Propylene carbonate can be used as a solvent in various reactions such as Biginelli reaction and Radziszewski reaction.

Application

Propylene carbonate can be used as a green solvent for the:
  • Extraction of polyhydroxy alkanoates from biomass.
  • Photodegradation of methylene blue.
  • Synthesis of glyceryl caffeate esters via lipase-catalyzed esterification.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

269.6 °F - closed cup

Flash Point(C)

132 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biginelli reaction using propylene carbonate as green solvent: An elegant methodology for the synthesis of dihydropyrimidinones and dihydropyrimidinthiones
Sikora A, et al.
Current Organic Synthesis, 14, 715-720 (2017)
Propylene carbonate as a solvent in the eco-friendly synthesis of highly substituted imidazoles through the Radziszewski reaction
A Hernandez M, et al.
Current Organic Synthesis, 13, 432-439 (2016)
Solubility and degradation of polyhydroxyalkanoate biopolymers in propylene carbonate
McChalicher C, et al.
AIChE Journal, 56, 1616-1625 (2010)
Enhanced methylene blue photodegradation with propylene carbonate as a solvent
Magalhaes P, et al.
Applied Surface Science, 458, 597-602 (2018)
Lipase-catalysed synthesis of mono-and di-acyl esters of glyceryl caffeate in propylene carbonate and their antioxidant properties in tuna oil
Cumming H, et al.
Journal of Biotechnology, 325, 217-225 (2021)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service