Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

32247

Sigma-Aldrich

Petroleum ether

puriss. p.a., reag. Ph. Eur., high boiling, bp 50-70 °C

Synonym(s):

Petroleum benzin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12191502
NACRES:
NA.21

Agency

USP/NF
reag. Ph. Eur.

vapor density

2.5 (vs air)

vapor pressure

25.8 psi

grade

puriss. p.a.

form

liquid

impurities

≤0.001% non-volatile matter
≤0.002% free acid (as CH3COOH)
≤0.005% S-compounds (as S)
≤0.01% benzene (GC)
≤0.01% water (Karl Fischer)

refractive index

n20/D 1.372-1.376

bp

50-70 °C

iodine value

≤0.3

density

0.661-0.664 g/mL at 20 °C

cation traces

Al: ≤0.5 mg/kg
B: ≤0.02 mg/kg
Ba: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.02 mg/kg
Sn: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg

Looking for similar products? Visit Product Comparison Guide

General description

hexane and low boiling point hydrogen treated naphtha

Application

Petroleum ether may be employed in the extraction of biologically active carotenoid pigments from total carotenoid pigments by adsorption. It may be employed in solvent reservoir during the separation of lipid mixtures by adsorption chromatography with silicic acid.

Other Notes

Pertains only in Germany: unversteuert

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Carc. 1B - Flam. Liq. 2 - Muta. 1B - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

<-40.0 °F - closed cup

Flash Point(C)

< -40 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ranganna S.
Handbook of Analysis and Quality Control for Fruit and Vegetable Products, 84-85 (1986)
The separation of complex lipide mixtures by the use of silicic acid chromatography.
J HIRSCH et al.
The Journal of biological chemistry, 233(2), 311-20 20-311-20 20 (1958-08-01)
S E Besra et al.
Journal of ethnopharmacology, 54(1), 1-6 (1996-10-01)
Pharmacological studies were conducted with the petroleum ether extract of leaves of the plant Litchi chinensis Gaertn. (Sapindaceae) on experimental animals. The extract was found to possess antiinflammatory, analgesic and antipyretic activity. Acute toxicity studies revealed that the extract, up

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service