Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.94520

Supelco

Petroleum benzine

parameter 40-60 °C, EMPLURA®

Synonym(s):

Petroleum ether

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12191502

product line

EMPLURA®

parameter

40-60 °C

dilution

(low-cost alternative, for basic applications)

storage temp.

20-25°C

Looking for similar products? Visit Product Comparison Guide

Related Categories

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

<-5.8 °F

Flash Point(C)

< -21 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service