Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

31119

Sigma-Aldrich

Ammonium sulfate

puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99%

Synonym(s):

Mascagnite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2SO4
CAS Number:
Molecular Weight:
132.14
EC Number:
MDL number:
UNSPSC Code:
12352300
eCl@ss:
38030408
PubChem Substance ID:
NACRES:
NA.21
form:
crystals

grade

ACS reagent
puriss. p.a.

Agency

USP/NF
reag. ISO
reag. Ph. Eur.

Assay

≥99%

form

crystals

impurities

≤0.005% in water insoluble matters
≤5 ppm heavy metals (by ICP-OES)

ign. residue

≤0.005% (as SO4)

loss

≤0.1% loss on drying, 110 °C

pH

5-6 (25 °C, 5%)

mp

>280 °C (dec.) (lit.)

density

1.77 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤3 mg/kg
nitrate (NO3-): ≤10 mg/kg
phosphate (PO43-): ≤5 mg/kg

cation traces

As: ≤0.2 mg/kg
Ca: ≤10 mg/kg
Cd: ≤1 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤2 mg/kg
Fe: ≤2 mg/kg
K: ≤50 mg/kg
Mg: ≤2 mg/kg
Mn: ≤5 mg/kg
Na: ≤20 mg/kg
Ni: ≤5 mg/kg
Pb: ≤2 mg/kg
Zn: ≤1 mg/kg

SMILES string

N.N.OS(O)(=O)=O

InChI

1S/2H3N.H2O4S/c;;1-5(2,3)4/h2*1H3;(H2,1,2,3,4)

InChI key

BFNBIHQBYMNNAN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ammonium sulfate is a versatile inorganic salt commonly used as an agriculture fertilizer as a nitrogen and sulfur source. It is also used as an extracting agent to extract metals such as iron, chromium, aluminum, and calcium. Furthermore, ammonium sulfate is employed in the purification of antibodies either from serum or ascitic fluids and in precipitating or fractionating proteins.

Application

Ammonium sulfatecan be used as a reagent in the palladium-catalyzed amination of aryl halidesto arylamines in the presence of sodium tert-butoxide. It can also beused as a catalyst in the hexamethyldisilazane-mediated reaction ofquinazolin-4(3H)-ones with primary amines to synthesize 4-aminoquinazolines.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ammonium sulfate preparation from phosphogypsum waste
Kandil, Abdel-Hakim K T, et al.
Journal of Radiation Research and Applied Sciences, 10, 24-33 (2017)
Extraction of magnesium from mine tailings for carbon dioxide mineralization: A preliminary study of the effect of ammonium sulfate to tailings ratio on products and yield.
Marjut M, et al.
Hydrometallurgy, 225, 106268-106268 (2024)
Antibody purification: ammonium sulfate fractionation or gel filtration
Ana Cristina G, et al.
Methods in Molecular Biology, 15-26 (2010)
D A DePasquale et al.
Applied and environmental microbiology, 56(12), 3711-3717 (1990-12-01)
In this study we examined the mechanism by which ammonium bicarbonate inhibits mycotoxigenic fungi. Elevated extracellular pH, alone, was not responsible for the antifungal activity. Although conidia of Penicillium griseofulvum and Fusarium graminearum had internal pH (pHi) values as high
Separation of proteins by ammonium sulfate gradient solubilization.
T P King
Biochemistry, 11(3), 367-371 (1972-02-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service