Skip to Content
MilliporeSigma
All Photos(3)

Documents

255572

Sigma-Aldrich

Potassium chlorate

ACS reagent, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KClO3
CAS Number:
Molecular Weight:
122.55
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NB.24

grade

ACS reagent

Quality Level

Assay

≥99.0%

form

crystalline powder

impurities

≤0.001% N compounds
≤0.005% insolubles

pH

5-6.5 (25 °C, 61.3 g/L)

mp

356 °C (lit.)

anion traces

bromate (BrO3-): ≤0.015%
chloride (Cl-): ≤0.001%
sulfate (SO42-): passes test

cation traces

Ca: ≤0.002%
Fe: ≤3 ppm
Mg: ≤0.002%
Na: ≤0.01%
heavy metals: ≤5 ppm (by ICP)

SMILES string

[K+].[O-]Cl(=O)=O

InChI

1S/ClHO3.K/c2-1(3)4;/h(H,2,3,4);/q;+1/p-1

InChI key

VKJKEPKFPUWCAS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Potassium chlorate (KClO3) is widely used in the chemical industry as an oxidizing agent. KCIO3 is used in the conversion of alcohols to corresponding aldehyde or ketone derivatives.

Application

Potassium chlorate is used:
  • In the sequential extraction (SEP) of selenium to analyze its isotopic composition from various Se pools.
  • In the preparation of graphite oxide (GO).

Pictograms

Flame over circleSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Ox. Sol. 1

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carbon fragments are ripped off from graphite oxide sheets during their thermal reduction.
Jankovsky O, et al.
New. J. Chem., 38(12), 5700-5705 (2014)
A noncovalent compatibilization approach to improve the filler dispersion and properties of polyethylene/graphene composites.
Vasileiou AA, et al.
ACS Applied Materials & Interfaces, 6(3), 1916-1925 (2014)
The Decomposition of Solid Potassium Chlorate by X Rays.
Heal HG.
Canadian Journal of Chemistry, 37(5), 979-987 (1959)
ESR Spectrum of O-3 Trapped in a Single Crystal of Potassium Chlorate.
Schlick S.
J. Chem. Phys. , 56, 654-661 (1972)
Daniel R Dreyer et al.
Chemical Society reviews, 39(1), 228-240 (2009-12-22)
The chemistry of graphene oxide is discussed in this critical review. Particular emphasis is directed toward the synthesis of graphene oxide, as well as its structure. Graphene oxide as a substrate for a variety of chemical transformations, including its reduction

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service