Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.06420

Supelco

Sodium chlorate

EMPLURA®

Synonym(s):

Sodium chlorate, Sodium Chlorate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
ClNaO3
CAS Number:
Molecular Weight:
106.44
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-887-4
NACRES:
NA.21
form:
solid

Quality Level

product line

EMPLURA®

Assay

≥98.0% (silver nitrate titration)

form

solid

potency

1200 mg/kg LD50, oral (Rat)
>10000 mg/kg LD50, skin (Rabbit)

pH

5-7 (20 °C, 50 g/L in H2O)

mp

255 °C (decomposition)

density

2.49 g/cm3 at 15 °C

bulk density

1500 kg/m3

anion traces

chloride (Cl-): ≤0.1%
sulfate (SO42-): ≤0.01%

cation traces

Fe: ≤0.005%
heavy metals (as Pb): ≤0.005%

storage temp.

2-30°C

SMILES string

[Na+].[Cl](=O)(=O)[O-]

InChI

1S/ClHO3.Na/c2-1(3)4;/h(H,2,3,4);/q;+1/p-1

InChI key

YZHUMGUJCQRKBT-UHFFFAOYSA-M

Application


  • Energy-efficient Chlorate Production through Adaptive Neuro-Fuzzy Modeling.: Recent advancements utilize an adaptive neuro-fuzzy inference system (ANFIS) to optimize sodium chlorate production by predicting cell pH. This approach significantly enhances energy efficiency, making it a pivotal development for industrial chemists focused on sustainable chemical manufacturing processes (Chemical Engineering Communications, 2020).

  • Solubility and Rheological Properties in Sodium Chlorate Systems.: In-depth studies on the polythermal solubility of sodium chlorate in ternary systems have revealed crucial data on its rheological properties. This research is vital for chemists engaged in solution chemistry and material science, facilitating the development of new materials and chemical processes (Russian Journal of Inorganic Chemistry, 2023).

Analysis Note

Assay (argentometric): ≥ 98.0 %
Chloride (Cl): ≤ 0.1 %
Sulfate (SO₄): ≤ 0.01 %
Heavy metals (as Pb): ≤ 0.005 %
Fe (Iron): ≤ 0.005 %

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Ox. Sol. 1

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service