Skip to Content
MilliporeSigma
All Photos(6)

Key Documents

241245

Sigma-Aldrich

Sodium acetate

ACS reagent, ≥99.0%

Synonym(s):

Acetic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COONa
CAS Number:
Molecular Weight:
82.03
Beilstein:
3595639
EC Number:
MDL number:
UNSPSC Code:
12352300
eCl@ss:
39021906
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

Assay

≥99.0%

form

solid

autoignition temp.

1112 °F

impurities

≤0.01% insolubles

loss

≤1.0% loss on drying

pH

7.0-9.2 (25 °C, 5%)

pKa 

4.76 (acetic acid)

mp

>300 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤0.002%
phosphate (PO43-): ≤0.001%
sulfate (SO42-): ≤0.003%

cation traces

Ca: ≤0.005%
Fe: ≤0.001%
Mg: ≤0.002%
heavy metals: ≤0.001% (by ICP-OES)

suitability

complies for IR spectroscopy

SMILES string

[Na+].CC([O-])=O

InChI

1S/C2H4O2.Na/c1-2(3)4;/h1H3,(H,3,4);/q;+1/p-1

InChI key

VMHLLURERBWHNL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Sodium acetate is a hydrophilic organic compound that can be prepared by treating acetic acid with sodium carbonate or sodium hydroxide. It acts as neutralizing agent, catalyst, food additive, and buffer to maintain a specific pH.

Application

Sodium acetate can be used as a base to synthesize:
  • Oxazolones via Erlenmeyer Plochl condensation reaction of aromatic aldehydes and hippuric acid.
  • Cinnamic acids via Perkin reaction between aromatic aldehydes and acid anhydrides.
A mixture of sodium acetate-acetic acid buffer can be employed in the polybromination of substituted imidazoles to synthesize polybrominated imidazoles.

It can also be used as a catalyst in tandem Knoevenagel-Michael multicomponent reactions.

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PREPARATION OF (S)-2-Allyl-2-methylcyclohexanone (Cyclohexanone, 2-methyl-2-(2-propen-1-yl)-, (2S)-).
Justin T Mohr et al.
Organic syntheses; an annual publication of satisfactory methods for the preparation of organic chemicals, 86, 194-211 (2009-01-01)
The liquid-chromatographic quantification of some synthetic colorants in soft drinks.
Trandafir I, et al.
Scientific Study and Research, 10(1), 73-82 (2009)
R E Kingston et al.
Current protocols in molecular biology, Chapter 4, Unit4-Unit4 (2008-02-12)
Three different methods for RNA preparation using guanidine are presented in this unit--a single-step isolation method employing liquid-phase separation to selectively extract total RNA from tissues and cultured cells and two methods that rely on a CsCl step gradient to
Listeria monocytogenes inhibition in refrigerated vacuum packaged turkey bologna by chemical additives.
Wederquist HJ, et al.
Journal of Food Science, 59(3), 498-500 (1994)
Eagleson M.
Concise Encyclopedia Chemistry, 995-995 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service