Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Y0001934

Deferoxamine for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Deferoxamine mesylate salt, DFOM, Deferoxamine methanesulfonate salt, Desferrioxamine mesylate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H48N6O8 · CH4O3S
CAS Number:
Molecular Weight:
656.79
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

deferoxamine

manufacturer/tradename

EDQM

format

neat

storage temp.

2-8°C

SMILES string

CS(O)(=O)=O.CC(=O)N(O)CCCCCNC(=O)CCC(=O)N(O)CCCCCNC(=O)CCC(=O)N(O)CCCCCN

InChI

1S/C25H48N6O8.CH4O3S/c1-21(32)29(37)18-9-3-6-16-27-22(33)12-14-25(36)31(39)20-10-4-7-17-28-23(34)11-13-24(35)30(38)19-8-2-5-15-26;1-5(2,3)4/h37-39H,2-20,26H2,1H3,(H,27,33)(H,28,34);1H3,(H,2,3,4)

InChI key

IDDIJAWJANBQLJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Deferoxamine for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

An iron chelator used often in the studies of cell proliferation and apoptosis. Also reported to induce p53.
An iron chelator used often in the studies of cell proliferation and apoptosis. Has been shown to have anti-proliferative effects on vascular smooth muscle cells in vitro and in vivo and to arrest cells in the G1 phase. Also reported to induce p53. Induces apoptosis in HL-60 cells by chelating iron. After 48 hrs treatment with 1μM deferoxamine, DNA fragmentation was apparent. Cells treated with 0.1 μM deferoxamine for as little as 24 hours were committed to apoptosis; by 48 hrs nuclear collapse was observed. In some studies it has been shown to have antioxidant properties and to protect cells against H2O2-induced damage.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service