Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Y0001575

Bupivacaine impurity F

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2,6-Dimethylaniline, 2,6-Xylidine, 2-Amino-1,3-dimethylbenzene, 2-Amino-m-xylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C6H3NH2
CAS Number:
Molecular Weight:
121.18
Beilstein:
636332
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

vapor pressure

<0.01 mmHg ( 20 °C)

API family

bupivacaine

manufacturer/tradename

EDQM

refractive index

n20/D 1.560 (lit.)

bp

214 °C/739 mmHg (lit.)

mp

10-12 °C (lit.)

density

0.984 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cc1cccc(C)c1N

InChI

1S/C8H11N/c1-6-4-3-5-7(2)8(6)9/h3-5H,9H2,1-2H3

InChI key

UFFBMTHBGFGIHF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Bupivacaine impurity F EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

195.8 °F - closed cup

Flash Point(C)

91 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

2,6-Dimethylaniline (2,6-xylidine).
IARC monographs on the evaluation of carcinogenic risks to humans, 57, 323-335 (1993-01-01)
Mai Shimizu et al.
Drug metabolism and disposition: the biological fate of chemicals, 42(7), 1103-1109 (2014-04-23)
Esterases catalyze the hydrolysis of therapeutic drugs containing esters or amides in their structures. Human carboxylesterase (CES) and arylacetamide deacetylase (AADAC) are the major enzymes that catalyze the hydrolysis of drugs in the liver. Characterization of the enzyme(s) responsible for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service