Skip to Content
MilliporeSigma
All Photos(1)

Documents

Y0001496

Cefoxitin sodium for peak identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Cefoxitin sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H16N3NaO7S2
CAS Number:
Molecular Weight:
449.43
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

cefoxitin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[Na+].CO[C@]2(NC(=O)Cc1cccs1)C3SCC(COC(N)=O)=C(N3C2=O)C([O-])=O

InChI

1S/C16H17N3O7S2.Na/c1-25-16(18-10(20)5-9-3-2-4-27-9)13(23)19-11(12(21)22)8(6-26-15(17)24)7-28-14(16)19;/h2-4,14H,5-7H2,1H3,(H2,17,24)(H,18,20)(H,21,22);/q;+1/p-1/t14-,16+;/m1./s1

InChI key

GNWUOVJNSFPWDD-XMZRARIVSA-M

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Cefoxitin sodium for peak identification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1B

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bioavailability and pharmacokinetics of cefoxitin sodium.
J J Schrogie et al.
The Journal of antimicrobial chemotherapy, 4(B), 69-78 (1978-07-01)
Clinical safety and tolerance of cefoxitin sodium: an overview.
C van Winzum
The Journal of antimicrobial chemotherapy, 4(B), 91-104 (1978-07-01)
Chun-Taek Oh et al.
The Journal of antimicrobial chemotherapy, 69(6), 1599-1607 (2014-02-13)
Mycobacterium abscessus is known to be the most drug-resistant Mycobacterium and accounts for ∼80% of pulmonary infections caused by rapidly growing mycobacteria. This study reports a new Drosophila melanogaster-M. abscessus infection model that can be used as an in vivo

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service