Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Y0001392

Olanzapine impurity A

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

5-Methyl-2-[(2-nitrophenyl)amino]-3-thiophenecarbonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H9N3O2S
CAS Number:
Molecular Weight:
259.28
Beilstein:
7485862
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

olanzapine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[s]1c(c(cc1C)C#N)Nc2c(cccc2)[N+](=O)[O-]

InChI

1S/C12H9N3O2S/c1-8-6-9(7-13)12(18-8)14-10-4-2-3-5-11(10)15(16)17/h2-6,14H,1H3

InChI key

NPXUFPFFHANGDL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Olanzapine impurity A EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Adrjanowicz et al.
The Journal of chemical physics, 136(23), 234509-234509 (2012-07-12)
The organic liquid ROY, i.e., 5-methyl-2-[(2-nitrophenyl)amino]-3-thiophenecarbonitrile, has been a subject of detailed study in the last few years. One interest in ROY lies in its polymorph-dependent fast crystal growth mode below and above the glass transition temperature. This growth mode
Christopher P Price et al.
Journal of the American Chemical Society, 127(15), 5512-5517 (2005-04-14)
The discovery and selective production of crystalline polymorphs, an outstanding problem in solid-state chemistry, is of great importance industrially in, for example, the manufacture of pharmaceuticals and pigments. Despite considerable efforts, no reliable method exists to produce all of the
Ying Diao et al.
Journal of the American Chemical Society, 134(1), 673-684 (2011-12-06)
Although nanoporous materials have been explored for controlling crystallization of polymorphs in recent years, polymorphism in confined environments is still poorly understood, particularly from a kinetic perspective, and the role of the local structure of the substrate has largely been
Conformational and color polymorphism of 5-methyl-2-[(2-nitrophenyl) amino]-3-thiophenecarbonitrile.
G A Stephenson et al.
Journal of pharmaceutical sciences, 84(11), 1385-1386 (1995-11-01)
Lian Yu
Accounts of chemical research, 43(9), 1257-1266 (2010-06-22)
Diamond and graphite are polymorphs of each other: they have the same composition but different structures and properties. Many other substances exhibit polymorphism: inorganic and organic, natural and manmade. Polymorphs are encountered in studies of crystallization, phase transition, materials synthesis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service