Skip to Content
MilliporeSigma

Skip To

1478323

USP

Olanzapine Related Compound B

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

5,10-Dihydro-2-methyl-4H-thieno[2,3-b][1,5]benzodiazepin-4-one, 2-Methyl-4,9-dihydro-3-thia-4,9-diazabenzo[f]azulene-10-one, Olanzapine lactam

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C12H10N2OS
CAS Number:
Molecular Weight:
230.29
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Beilstein/REAXYS Number:
8262494
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

olanzapine

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[S]1=C2Nc3c(cccc3)NC(=O)C2=CC=1C

InChI

1S/C12H10N2OS/c1-7-6-8-11(15)13-9-4-2-3-5-10(9)14-12(8)16-7/h2-6,14H,1H3,(H,13,15)

InChI key

LVTDAJJOMLNXFS-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR25371478312PHR2538
grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

USP

manufacturer/tradename

-

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical small molecule

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical small molecule

format

neat

format

-

format

neat

format

-

API family

olanzapine

API family

olanzapine

API family

olanzapine

API family

olanzapine


Still not finding the right product?

Explore all of our products under Olanzapine Related Compound B


Hazard Classifications

STOT SE 3

pictograms

Exclamation mark

signalword

Warning

hcodes

pcodes

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters