Skip to Content
MilliporeSigma
All Photos(1)

Documents

W200506

Sigma-Aldrich

Acetanisole

≥98%, FCC, FG

Synonym(s):

4′-Methoxyacetophenone, p-Acetanisole, 4-Acetylanisole

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4COCH3
CAS Number:
Molecular Weight:
150.17
FEMA Number:
2005
Beilstein:
742313
EC Number:
Council of Europe no.:
570
MDL number:
UNSPSC Code:
12164502
eCl@ss:
39023808
PubChem Substance ID:
Flavis number:
7.038
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥98%

bp

152-154 °C/26 mmHg (lit.)

mp

36-38 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

anise; floral

SMILES string

COc1ccc(cc1)C(C)=O

InChI

1S/C9H10O2/c1-7(10)8-3-5-9(11-2)6-4-8/h3-6H,1-2H3

InChI key

NTPLXRHDUXRPNE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acetanisole is an aromatic ketone that occurs naturally in anise seeds. It is mainly used as a flavoring agent in ice-creams, non-alcoholic beverages and baked goods.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

280.4 °F - open cup

Flash Point(C)

138 °C - open cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Burdock GA
Encyclopedia of Food and Color Additives, 1, 11-12 (1997)
Fenaroli G
Fenaroli's Handbook of Flavor Ingredients, 5-5 (1971)
Burdock GA
Encyclopedia of Food and Color Additives, 1, 11-12 (1997)
Wen-Yong Lou et al.
Journal of biotechnology, 143(3), 190-197 (2009-07-21)
The biocatalytic enantioselective reduction of 4'-methoxyacetophenone to (S)-1-(4-methoxyphenyl)ethanol was successfully conducted in a hydrophilic IL-containing co-solvent system using immobilized Rhodotorula sp. AS2.2241 cells. Of all the tested ILs, the best results were observed with the novel IL 1-(2'-hydroxy)ethyl-3-methylimidazolium nitrate (C(2)OHMIM.NO(3))
[Toxicological characteristics of acetylanisole].
M I Makaruk et al.
Gigiena i sanitariia, (4)(4), 86-87 (1985-04-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service