Skip to Content
MilliporeSigma
L-Tryptophan certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

L-Tryptophan

L-Tryptophan certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
(S)-2-Amino-3-(3-indolyl)propionic acid, L-α-Amino-3-indolepropionic acid, L-Tryptophan
Empirical Formula (Hill Notation):
C11H12N2O2
CAS Number:
Molecular Weight:
204.23
EC Number:

Products

Product Category
Brand
Color
Feature
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Analyte Suitability
Quality Segment
Application
Optical Isomer
Amino Acid
Reference Material Type
Special Grade
Technique