Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SMB01351

Sigma-Aldrich

γ-Glutamylphenylalanine

≥95% (HPLC)

Synonym(s):

γ-Glu-Phe, γ-Glutamyl-L-phenylalanine, γ-Glutamylphenylalanine, L-γ-Glutamyl-L-phenylalanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H18N2O5
CAS Number:
Molecular Weight:
294.30
MDL number:
UNSPSC Code:
12352209

Quality Level

Assay

≥95% (HPLC)

form

powder or crystals

mp

194—197 °C

solubility

water: slightly soluble

cation traces

C: 54.3—60%

storage temp.

2-8°C

InChI

1S/C14H18N2O5/c15-10(13(18)19)6-7-12(17)16-11(14(20)21)8-9-4-2-1-3-5-9/h1-5,10-11H,6-8,15H2,(H,16,17)(H,18,19)(H,20,21)/t10-,11-/m0/s1

InChI key

XHHOHZPNYFQJKL-QWRGUYRKSA-N

General description

γ-Glutamylphenylalanine is a dipeptide composed of gamma-glutamate and phenylalanine, and is a proteolytic breakdown product of larger proteins. It is believed to be formed by transpeptidation between glutathione and the corresponding amino acid, catalyzed by gamma-glutamyl transpeptidase. Some dipeptides are known to have physiological or cell-signaling effects although most are simply short-lived intermediates on their way to specific amino acid degradation pathways following further proteolysis. It can be detected in the urine of patients with phenylketonuria, which is a human genetic disorder due to the lack of phenylalanine hydroxylase to metabolize phenylalanine to tyrosine. gamma-Glutamylphenylalanine is found in adzuki bean, onion, garlic, and soybean.

Application

γ-Glutamylphenylalanine can be used in biochemical and metabolomics research applications

Features and Benefits

  • Suitable for Metabolomics and Biochemical research
  • High-quality compound suitable for multiple research applications

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service