Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SMB01326

Sigma-Aldrich

5-Hydroxytryptophol

Synonym(s):

5-Hydroxy-1H-indole-3-ethanol, 5-Hydroxy-3-(2-hydroxyethyl)indole, 2-(5-Hydroxy-3-indolyl)ethyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11NO2
CAS Number:
Molecular Weight:
177.20
MDL number:
UNSPSC Code:
12352205
NACRES:
NA.77

Assay

99% (HPLC)

Quality Level

form

solid

storage temp.

2-8°C

SMILES string

OCCc1c[nH]c2ccc(O)cc12

InChI

1S/C10H11NO2/c12-4-3-7-6-11-10-2-1-8(13)5-9(7)10/h1-2,5-6,11-13H,3-4H2

InChI key

KQROHCSYOGBQGJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

5-Hydroxytryptophol is metabolite of serotonin that is excreted primarily as the glucuronide conjugate in human urine. 5-Hydroxytryptophol makes up 35% of the excreted serotonin metabolites in rats on average and 10 to 20% in several other species. Human UDP-glucuronosyltransferase 1A6 (UGT1A6) plays a predominant role in the glucuronidation of 5-hydroxytryptophol by human liver microsomes.

Application

Metabolomics research

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service