Skip to Content
MilliporeSigma

C3633

Cystathionine

≥90% (HPLC)

Synonym(s):

DL,DL-allo-Cystathionine, S-(2-Amino-2-carboxyethyl)homocysteine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
HOOCCH(NH2)CH2CH2SCH2CH(NH2)COOH
CAS Number:
Molecular Weight:
222.26
NACRES:
NA.26
PubChem Substance ID:
UNSPSC Code:
12352209
EC Number:
208-613-7
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Cystathionine, ≥90% (HPLC)

Quality Segment

assay

≥90% (HPLC)

form

powder

color

white to off-white

mp

281 °C (dec.)

storage temp.

2-8°C

SMILES string

NC(CCSCC(N)C(O)=O)C(O)=O

InChI

1S/C7H14N2O4S/c8-4(6(10)11)1-2-14-3-5(9)7(12)13/h4-5H,1-3,8-9H2,(H,10,11)(H,12,13)

InChI key

ILRYLPWNYFXEMH-UHFFFAOYSA-N

Gene Information

human ... BHMT(635)

Biochem/physiol Actions

Cystathionine is an intermediate in the biosynthesis of the amino acid cysteine. Cystathionine is used as a substrate to differentiate and analyze cystathionine gamma-lyase(s).

Preparation Note

Prepared by method of Stekol.

Other Notes

A mixture of 4 stereoisomers; L-, D-, L-allo- and D-allo-cystathionine.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library