Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SMB00978

Sigma-Aldrich

L-Hercynine

≥95% (HPLC)

Synonym(s):

α-N,N,N-Trimethylhistidine, N-α-trimethyl histidine, Histidine-betaine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H15N3O2
CAS Number:
Molecular Weight:
197.23
UNSPSC Code:
41116105
NACRES:
NA.26

Quality Level

Assay

≥95% (HPLC)

form

solid

storage temp.

2-8°C

SMILES string

C[N+](C)(C)[C@H](C([O-])=O)CC1=CN=CN1

InChI

1S/C9H15N3O2/c1-12(2,3)8(9(13)14)4-7-5-10-6-11-7/h5-6,8H,4H2,1-3H3,(H-,10,11,13,14)/t8-/m0/s1

InChI key

GPPYTCRVKHULJH-QMMMGPOBSA-N

General description

Hercynine is the precursor and redox metabolite of ergothioneine, which regulates microbial physiology and enables the survival of microbes under stressful conditions encountered in their natural environments. Ergothioneine is primarily present as a thione physiological pH, which makes it resistant to autoxidation. Ergothioneine enables pathogenic microbes, such as Mycobacterium tuberculosis (Mtb) to withstand hostile environments within the host to establish infection.

Application

Hercynine is a versatile molecule and it finds application in biochemical and metabolomics research.

Features and Benefits

  • Can be used in Metabolomics and Biochemical research
  • High-quality compound suitable for multiple research applications

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katarina Valachova et al.
Polymers, 13(1) (2021-01-02)
A high-molecular weight hyaluronan is oxidatively degraded by Cu(II) ions and ascorbate-the so called Weissberger biogenic oxidative system-which is one of the most potent generators of reactive oxygen species, namely •OH radicals. Ergothioneine, hercynine, or histidine were loaded into chitosan/hyaluronan
Salvatore Sotgia et al.
Antioxidants (Basel, Switzerland), 9(9) (2020-09-17)
The dependence of a stallion's spermatozoa on oxidative phosphorylation for energy requirements results in an unconventional relationship between reactive oxygen species (ROS) production and fertility. In such a scenario, antioxidant activity must be finely controlled and not affect the essential
Tomoyuki Kamide et al.
Journal of agricultural and food chemistry, 68(23), 6390-6394 (2020-05-22)
We previously constructed a heterologous production system for ergothioneine (ERG) in Escherichia coli using five ERG biosynthesis genes (egtABCDE) from Mycobacterium smegmatis. However, significant amounts of hercynine (HER), an intermediate of ERG, as ERG were accumulated, suggesting that the reaction

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service