Skip to Content
MilliporeSigma
All Photos(1)

Documents

SMB00917

Sigma-Aldrich

Deuterated Amino Acid Standard Mixture

suitable for mass spectrometry (MS)

Synonym(s):

Deuterated Amino Acid Standard Mixture, AA mixture, Amino Acids mixture

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.21

form

liquid

Quality Level

IVD

for in vitro diagnostic use

technique(s)

mass spectrometry (MS): suitable

application(s)

metabolomics

shipped in

wet ice

storage temp.

2-8°C

Application

The Deuterated Amino Acid Standard Mixture is formulated for use as a concentration standard for high throughput LC/MS or GC-MS metabolomic analysis.

Features and Benefits

  • All deuterated amino acids are dissolved in water and ready to use. Total vial volume of 10 ml.
  • Ready to use deuterated internal standards.
  • Ideal for polar metabolites for HILIC chromatography.

Analyte

Description

    Glycine (D5)
    L-Histidine (D5)
    L-Serine (2,3,3-D3)
    L-Alanine (2,3,3,3-D4)
    L-Isoleucine (D10)
    L-Threonine (D5)
    L-Arginine (D7)
    L-Leucine (D10)
    L-Tryptophan (D8)
    L-Asparagine (2,3,3-D3)
    L-Lysine (D8)
    L-Tyrosine (D7)
    L-Aspartic acid (2,3,3-D3)
    L-Methionine (D8)
    L-Valine (D8)
    L-Cystine (3,3,3′,3′-D4)
    L-Ornithine (5,5-D2)
    PI-Methyl-L-Histidine (D3)
    L-Glutamic acid (2,3,3,4,4-D5)
    L-Phenylalanine (D8)
    TAU-Methyl-L-Histidine (D3)
    L-Glutamine (2,3,3,4,4-D5)
    L-Proline (D7)
    DL-Homocysteine (3,3,4,4-D4)
    See All (24)

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oliver Fiehn
Current protocols in molecular biology, 114, 30-30 (2016-04-03)
Gas chromatography-mass spectrometry (GC-MS)-based metabolomics is ideal for identifying and quantitating small-molecule metabolites (<650 Da), including small acids, alcohols, hydroxyl acids, amino acids, sugars, fatty acids, sterols, catecholamines, drugs, and toxins, often using chemical derivatization to make these compounds sufficiently
Dinesh K Barupal et al.
Metabolites, 9(5) (2019-05-28)
Mouse knockouts facilitate the study ofgene functions. Often, multiple abnormal phenotypes are induced when a gene is inactivated. The International Mouse Phenotyping Consortium (IMPC) has generated thousands of mouse knockouts and catalogued their phenotype data. We have acquired metabolomics data

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service