Skip to Content
MilliporeSigma
All Photos(1)

Documents

S8394

Supelco

Silica gel Inorganic Sorbent

550 mesh, packet of 5 g desiccant

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
23201100
PubChem Substance ID:
NACRES:
SB.52

product name

Silica gel, packet of 5 g desiccant

grade

reagent grade

Quality Level

form

powder

packaging

packet of 5 g desiccant

technique(s)

LPLC: suitable

surface area

500 m2/g , approx.

matrix

Silica

matrix active group

silica

particle size

25 μm

pore size

60 Å mean pore size

bp

2230 °C

mp

>1600 °C

separation technique

hydrophilic interaction (HILIC)

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Flexible and Compressible Nanostructure-Assembled Aramid Nanofiber/Silica Composites Aerogel: This study explores the development of flexible and compressible silica-based aerogels, highlighting their potential in lightweight construction and thermal insulation applications, particularly useful for research in material science and industrial manufacturing (Zhang et al., 2024).
  • Factors Affecting Silica/Cellulose Nanocomposite Prepared via the Sol-Gel Technique: A Review: This review discusses the synthesis of silica/cellulose nanocomposites using the sol-gel method, emphasizing the impact of various factors on the material properties, relevant for researchers in materials science and environmental applications (Shange et al., 2024).
  • Spray-dried ternary bioactive glass microspheres: direct and indirect structural effects of copper-doping on acellular degradation behavior: Investigating the structural properties of copper-doped silica bioactive glass, this paper focuses on their applications in biomedicine and drug delivery systems, particularly in bone regeneration and tissue engineering (Vecchio et al., 2024).
  • Isolation, characterization, identification and quantification of 6-F oxyphenisatin dipropionate, a novel illegal additive, from a fruit-flavored jelly: Demonstrating the use of silica gel in chromatographic techniques for detecting illegal additives in food products, this study is vital for food safety and environmental testing laboratories (Xia et al., 2024).

Packaging

Porous 1 × 23/4 in. packet

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service