Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

WHA10548239

Whatman® Desiccant Pack

packet of 1 g, pack of 100 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104510
NACRES:
NB.24

packaging

packet of 1 g
pack of 100 ea

manufacturer/tradename

Whatman 10548239

General description

1 g packs of desiccant ensure that DNA sampling cards remain dry during transport or storage, 100/pk.

Features and Benefits

  • High vapor transmission rate
  • Substrate is made with Tyvek® material
  • Non-dusting, strong and hard to tear

Legal Information

Tyvek is a registered trademark of E. I. du Pont de Nemours and Company
Whatman is a registered trademark of Cytiva

Storage Class Code

13 - Non Combustible Solids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service