Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

S1429

Sigma-Aldrich

Sodium acetate

meets USP testing specifications, anhydrous

Synonym(s):

Acetic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COONa
CAS Number:
Molecular Weight:
82.03
Beilstein:
3595639
EC Number:
MDL number:
UNSPSC Code:
12352300
eCl@ss:
39021906
PubChem Substance ID:
NACRES:
NA.21

grade

anhydrous

Quality Level

Agency

USP/NF
meets USP testing specifications

Assay

99-101% dry basis

form

solid

autoignition temp.

1112 °F

pH

8.5-9.9 (25 °C, 246 g/L)

pKa 

4.76 (acetic acid)

mp

>300 °C (dec.) (lit.)

solubility

soluble 246 g/L at 20 °C (completely)

application(s)

pharmaceutical (small molecule)

SMILES string

[Na+].CC([O-])=O

InChI

1S/C2H4O2.Na/c1-2(3)4;/h1H3,(H,3,4);/q;+1/p-1

InChI key

VMHLLURERBWHNL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium acetate (SA) exhibits antimicrobial and antioxidant properties. It is used as an exchange-fluid buffer in hemodialysis. SA acts as an a alkalinizing agent and its continuous intravenous (IV) infusion causes salicylate intoxication.

Application

Sodium acetate (SA) has been used to increase hepatic acetyl- coenzyme A in caloric restricted rats.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sodium Acetate as an Alkalinizing Agent for Salicylate Intoxication: A Case Report
Groke SF, et al.
Journal of Clinical Toxicology (2015)
Enhancement of in-vitro human interleukin-1 production by sodium acetate
Bingel M, et al.
Lancet, 329(8523), 14-16 (1987)
Antimicrobial and antioxidant effects of sodium acetate, sodium lactate, and sodium citrate in refrigerated sliced salmon
Sallam KI, et al.
Food Control, 18(5), 566-575 (2007)
Mechanisms by which a very-low-calorie diet reverses hyperglycemia in a rat model of type 2 diabetes
Perry RJ, et al.
Cell Metabolism, 27(1), 210-217 (2018)
Graham R Scott et al.
Molecular biology and evolution, 32(8), 1962-1976 (2015-04-09)
At high-altitude, small mammals are faced with the energetic challenge of sustaining thermogenesis and aerobic exercise in spite of the reduced O2 availability. Under conditions of hypoxic cold stress, metabolic demands of shivering thermogenesis and locomotion may require enhancements in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service