Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

PPB011

Sigma-Aldrich

Tris Buffered Saline

powder, pH 7.6, pHast Pack

Synonym(s):

TBS

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12161700
NACRES:
NA.25

Product Name

Tris Buffered Saline, pH 7.6, pHast Pack, powder

product line

pHast Pack

Quality Level

form

powder

storage condition

(Tightly closed. Dry)

impurities

≤5 ppm Heavy Metals (lead)

color

white

pH

7.6

solubility

soluble, clear, colorless

cation traces

Fe: ~10 ppm

foreign activity

DNase, none detected
Endonuclease, none detected
Exonuclease, none detected
NICKase, none detected
Protease, none detected
RNase, none detected

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

General description

Tris Buffered Saline, or TBS, pH 7.6 is a commonly used wash buffer that is isotonic and non-toxic and ideal for use in cell culture and electrophoresis applications.

Application

Tris-buffered saline can be used as a wash buffer for:
  • Enzyme Linked Immunosorbent assay (ELISA)
  • Western Blot and other protein applications
  • Suitable for use in cell culture (recommend to sterile filter)
  • Immunohistochemistry
  • Sample preparation
  • Resuspension and sonication of cells

Features and Benefits

  • Ready for use, just dissolve in ultrapure water
  • No calculations, measuring, and pH adjustments needed
  • Save time and minimize effort
  • Biological tests: free of DNase, RNase, Protease, and Nickase
  • Chemical tests: Iron ≤10 ppm, Lead ≤5 ppm

Packaging

Foil pouches

Reconstitution

Contents of one pouch, when dissolved in 500 mL of ultrapure water, will yield a 1x solution containing 50 mM Tris, 138 mM NaCl, 2.7 mM KCl, pH 7.6 at 25 °C. Contents tested to be DNAse, RNAse, Protease, and Nickase free. This product is suitable for use in cell culture applications.

Storage and Stability

Store at room temperature. Product may naturally agglomerate but can be simply broken up within the pouch prior to use.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Legal Information

pHast Pack is a trademark of Sigma-Aldrich Co. LLC

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service