Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PHR8919

Supelco

Dioxindolyl-L-alanine

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

(2S)-2-Amino-3-[(3RS)-3-hydroxy-2-oxo-2,3-dihydro-1H-indol-3-yl]propanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12N2O4
CAS Number:
Molecular Weight:
236.22
UNSPSC Code:
12352209
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

API family

tryptophan

form

solid

packaging

pkg of 100 mg

application(s)

pharmaceutical small molecule

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service