Skip to Content
MilliporeSigma

PHR8908

N-Formylkynurenine

certified reference material, pharmaceutical secondary standard

Synonym(s):

(2S)-2-Amino-4-[2-(formylamino)phenyl]-4-oxobutanoic acid, (2S)-2-Amino-4-[2-(formylamino)phenyl]-4-oxobutanoic acid, Tryptophan Impurity E (Eur. Phr.)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C11H12N2O4
CAS Number:
Molecular Weight:
236.22
UNSPSC Code:
41116107
NACRES:
NA.24
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

grade

certified reference material, pharmaceutical secondary standard

API family

tryptophan

form

solid

packaging

pkg of 100 mg

application(s)

pharmaceutical small molecule

SMILES string

[N+H3][C@@H](CC(=O)c1c(cccc1)NC=O)C(=O)[O-]

InChI

1S/C11H12N2O4/c12-8(11(16)17)5-10(15)7-3-1-2-4-9(7)13-6-14/h1-4,6,8H,5,12H2,(H,13,14)(H,16,17)/t8-/m0/s1

InChI key

BYHJHXPTQMMKCA-QMMMGPOBSA-N

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP and EP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library