Skip to Content
MilliporeSigma

PHR2201

Trans-Resveratrol

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Resveratrol, 3,4′,5-Trihydroxy-trans-stilbene, 5-[(1E)-2-(4-Hydroxyphenyl)ethenyl]-1,3-benzenediol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C14H12O3
CAS Number:
Molecular Weight:
228.24
UNSPSC Code:
41116107
NACRES:
NA.24
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

grade

certified reference material, pharmaceutical secondary standard

agency

traceable to Ph. Eur. Y0001194, traceable to USP 1602105

API family

resveratrol

CofA

current certificate can be downloaded

packaging

pkg of 200 mg

application(s)

pharmaceutical small molecule

format

neat

storage temp.

2-8°C

SMILES string

Oc1ccc(cc1)\C=C\c2cc(O)cc(O)c2

InChI

1S/C14H12O3/c15-12-5-3-10(4-6-12)1-2-11-7-13(16)9-14(17)8-11/h1-9,15-17H/b2-1+

InChI key

LUKBXSAWLPMMSZ-OWOJBTEDSA-N

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Application

Selective inhibitor of COX−1.

Analysis Note

These secondary standards offer multi-traceability to the USP and EP primary standards, where they are available.

Other Notes

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
To see an example of a Certificate of Analysis for this material enter LRAC2237 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.


Still not finding the right product?

Explore all of our products under Trans-Resveratrol


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library