Skip to Content
MilliporeSigma
trans-Resveratrol phyproof® Reference Substance

trans-Resveratrol

trans-Resveratrol phyproof® Reference Substance
Synonym(s):
Resveratrol, 3,4′,5-Trihydroxy-trans-stilbene, 5-[(1E)-2-(4-Hydroxyphenyl)ethenyl]-1,3-benzenediol
Empirical Formula (Hill Notation):
C14H12O3
CAS Number:
Molecular Weight:
228.24

Products

Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade