Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PHR2005

Supelco

Allopurinol Related Compound A

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

3-Amino-4-pyrazolecarboxamide hemisulfate salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C4H6N4O)2 · H2SO4
CAS Number:
Molecular Weight:
350.31
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 831
traceable to Ph. Eur. A0350010
traceable to USP 1013024

API family

allopurinol

form

solid

CofA

current certificate can be downloaded

packaging

pkg of 50 mg

mp

224 °C (dec.) (lit.)

application(s)

pharmaceutical

storage temp.

2-8°C

SMILES string

OS(O)(=O)=O.NC(=O)c1c[nH]nc1N.NC(=O)c2c[nH]nc2N

InChI

1S/2C4H6N4O.H2O4S/c2*5-3-2(4(6)9)1-7-8-3;1-5(2,3)4/h2*1H,(H2,6,9)(H3,5,7,8);(H2,1,2,3,4)

InChI key

UMPKASYMNORSRO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAB6412 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service