Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
grade
pharmaceutical primary standard
API family
allopurinol
manufacturer/tradename
EDQM
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Packaging
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| description European Pharmacopoeia (EP) Reference Standard | description European Pharmacopoeia (EP) Reference Standard | description European Pharmacopoeia (EP) Reference Standard | description European Pharmacopoeia (EP) Reference Standard |
| manufacturer/tradename EDQM | manufacturer/tradename EDQM | manufacturer/tradename EDQM | manufacturer/tradename EDQM |
| grade pharmaceutical primary standard | grade pharmaceutical primary standard | grade pharmaceutical primary standard | grade pharmaceutical primary standard |
| format neat | format neat | format neat | format neat |
| application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) |
| API family allopurinol | API family allopurinol | API family allopurinol | API family allopurinol |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C |
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


