Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PHR1737

Supelco

Colistimethate Sodium

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Colistimethate sodium, CMS, Colistin sulphomethate sodium

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. C2600000
traceable to USP 1147009

API family

colistimethate

CofA

current certificate can be downloaded

packaging

pkg of 500 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

-10 to -25°C

SMILES string

[Na+].[Na+].[Na+].[Na+].[Na+].[S](=O)(=O)([O-])CNCCC1NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(CCNC(=O)C(NC1=O)C(O)C)NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)CCCCC(CC)C)CCNC[S](=O)(=O)[O-])C(O)C)CCNC[S](=O)(=O)[O-])CCNC[S](=O)(=O)[O-])CC(C)C)CC(C)C)CCNC[S](=O)(=O)[O-]

InChI

1S/C58H110N16O28S5.5Na/c1-9-35(6)12-10-11-13-46(77)65-38(14-20-59-28-103(88,89)90)53(82)74-48(37(8)76)58(87)70-41(17-23-62-31-106(97,98)99)50(79)68-43-19-25-64-57(86)47(36(7)75)73-54(83)42(18-24-63-32-107(100,101)102)67-49(78)39(15-21-60-29-104(91,92)93)69-55(84)44(26-33(2)3)72-56(85)45(27-34(4)5)71-52(81)40(66-51(43)80)16-22-61-30-105(94,95)96;;;;;/h33-45,47-48,59-63,75-76H,9-32H2,1-8H3,(H,64,86)(H,65,77)(H,66,80)(H,67,78)(H,68,79)(H,69,84)(H,70,87)(H,71,81)(H,72,85)(H,73,83)(H,74,82)(H,88,89,90)(H,91,92,93)(H,94,95,96)(H,97,98,99)(H,100,101,102);;;;;/q;5*+1/p-5

InChI key

IQWHCHZFYPIVRV-UHFFFAOYSA-I

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Colistimethate Sodium belongs to the class of polymyxin antibiotics that acts by inhibiting the Gram negative bacteria. It is a prodrug that can hydrolyze to colistin in vivo or in vitro. It is employed as a last resort antibiotic to treat Pseudomonas aeruginosa infection in cystic fibrosis patients.

Application

Colistimethate Sodium may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by high performance liquid chromatography (HPLC).
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA8043 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A simple method for assaying colistimethate sodium in pharmaceutical aerosol samples using high performance liquid chromatography
Metcalf AP, et al.
Journal of Pharmaceutical and Biomedical Analysis, 142, 15-18 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service